Assay Details
| Gene Symbol: PWWP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075856.2 | NP_001069324.2 | 135 | 107 | ||
| GenBank mRNA | BC118135.1 | - | 136 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118135.1 transcripts.
Set Membership: |
|

PWWP2B
Genomic Map