Assay Details
| Gene Symbol: QRICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098015.2 | NP_001091484.1 | 7 - 8 | 2069 | 64 | |
| GenBank mRNA | BC119972.1 | - | 5 - 6 | 1465 | 64 | |
| BC140621.1 | - | 7 - 8 | 2070 | 64 | ||
More Information
Set Membership: |
|

QRICH1
Genomic Map