Assay Details
| Gene Symbol: TMEM88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098378.2 | NP_001091848.1 | 1 - 2 | 219 | 70 | |
| GenBank mRNA | BC140617.1 | - | 1 - 2 | 220 | 70 | |
More Information
Set Membership: |
|

TMEM88
Genomic Map