Assay Details
| Gene Symbol: MED26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098888.1 | NP_001092358.1 | 1 - 2 | 330 | 51 | |
| GenBank mRNA | BC142325.1 | - | 1 - 2 | 330 | 51 | |
More Information
Set Membership: |
|

MED26
Genomic Map