Assay Details
| Gene Symbol: SUDS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098891.1 | NP_001092361.1 | 11 - 12 | 1036 | 60 | |
| GenBank mRNA | BC146027.1 | - | 11 - 12 | 1036 | 60 | |
More Information
Set Membership: |
|

SUDS3
Genomic Map