Assay Details
| Gene Symbol: DTYMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098893.1 | NP_001092363.1 | 2 - 3 | 260 | 74 | |
| GenBank mRNA | BC142258.1 | - | 2 - 3 | 260 | 74 | |
More Information
Set Membership: |
|

DTYMK
Genomic Map