Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098895.2 | NP_001092365.1 | 2 - 3 | 195 | 59 | |
| GenBank mRNA | BC146143.1 | - | 2 - 3 | 196 | 59 | |
More Information
Set Membership: |
|

SLC25A30
Genomic Map