Assay Details
| Gene Symbol: AAGAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098896.1 | NP_001092366.1 | 9 - 10 | 897 | 88 | |
| GenBank mRNA | BC142460.1 | - | 9 - 10 | 897 | 88 | |
More Information
Set Membership: |
|

AAGAB
Genomic Map