Assay Details
| Gene Symbol: PAQR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098897.1 | NP_001092367.1 | 1 - 2 | 500 | 61 | |
| GenBank mRNA | BC146103.1 | - | 1 - 2 | 500 | 61 | |
More Information
Set Membership: |
|

PAQR4
Genomic Map