Assay Details
| Gene Symbol: FAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098912.1 | NP_001092382.1 | 2 - 3 | 210 | 59 | |
| GenBank mRNA | BC142487.1 | - | 2 - 3 | 210 | 59 | |
| BC151490.1 | - | 2 - 3 | 224 | 59 | ||
| BC151824.1 | - | 2 - 3 | 210 | 59 | ||
More Information
Set Membership: |
|

FAH
Genomic Map