Assay Details
| Gene Symbol: SPATA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098928.2 | NP_001092398.1 | 6 - 7 | 934 | 77 | |
| GenBank mRNA | BC146052.1 | - | 6 - 7 | 935 | 77 | |
More Information
Set Membership: |
|

SPATA7
Genomic Map