Assay Details
| Gene Symbol: HMGN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098945.1 | NP_001092415.1 | 1 - 2 | 118 | 91 | |
| GenBank mRNA | BC142241.1 | - | 1 - 2 | 118 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC142241.1, NM_001098945.1 transcripts.
Set Membership: |
|

HMGN2
Genomic Map