Assay Details
| Gene Symbol: SNRPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098948.2 | NP_001092418.1 | 4 - 5 | 389 | 69 | |
| GenBank mRNA | BC146154.1 | - | 4 - 5 | 391 | 69 | |
More Information
Set Membership: |
|

SNRPA1
Genomic Map