Assay Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098962.2 | NP_001092432.1 | 3 - 4 | 481 | 60 | |
| GenBank mRNA | BC142025.1 | - | 3 - 4 | 482 | 60 | |
More Information
Set Membership: |
|

CCDC43
Genomic Map