Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099013.1 | NP_001092483.1 | 16 - 17 | 1886 | 81 | |
| GenBank mRNA | BC142486.1 | - | 16 - 17 | 1886 | 81 | |
More Information
Set Membership: |
|

DNAI3
Genomic Map