Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099013.1 | NP_001092483.1 | 9 - 10 | 1145 | 89 | |
| GenBank mRNA | BC142486.1 | - | 9 - 10 | 1145 | 89 | |
More Information
Set Membership: |
|

DNAI3
Genomic Map