Assay Details
| Gene Symbol: ABHD17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099014.1 | NP_001092484.1 | 2 - 3 | 827 | 65 | |
| GenBank mRNA | BC142452.1 | - | 2 - 3 | 827 | 65 | |
More Information
Set Membership: |
|

ABHD17C
Genomic Map