Assay Details
| Gene Symbol: POC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099031.1 | NP_001092501.1 | 11 - 12 | 1360 | 81 | |
| GenBank mRNA | BC142494.1 | - | 11 - 12 | 1360 | 81 | |
More Information
Set Membership: |
|

POC1B
Genomic Map