Assay Details
| Gene Symbol: CD200R1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099374.2 | NP_001092844.1 | 6 - 7 | 1071 | 81 | |
| GenBank mRNA | BC140695.1 | - | 6 - 7 | 1072 | 81 | |
More Information
Set Membership: |
|

CD200R1L
Genomic Map