Assay Details
| Gene Symbol: LOC510193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100333.1 | NP_001093803.1 | 2 - 3 | 150 | 64 | |
| GenBank mRNA | BC149201.1 | - | 2 - 3 | 150 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149201.1, NM_001100333.1 transcripts.
Set Membership: |
|

LOC510193
Genomic Map