Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100344.1 | NP_001093814.1 | 2 - 3 | 701 | 83 | |
| GenBank mRNA | BC149816.1 | - | 2 - 3 | 701 | 83 | |
More Information
Set Membership: |
|

ZKSCAN8
Genomic Map