Assay Details
| Gene Symbol: MIOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101065.1 | NP_001094535.1 | 8 - 9 | 652 | 72 | |
| GenBank mRNA | BC151511.1 | - | 8 - 9 | 652 | 72 | |
More Information
Set Membership: |
|

MIOX
Genomic Map