Assay Details
| Gene Symbol: RUVBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101076.1 | NP_001094546.1 | 9 - 10 | 1154 | 65 | |
| GenBank mRNA | BC151551.1 | - | 9 - 10 | 1154 | 65 | |
More Information
Set Membership: |
|

RUVBL1
Genomic Map