Assay Details
| Gene Symbol: ADAMTS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101080.1 | NP_001094550.1 | 3 - 4 | 1678 | 73 | |
| GenBank mRNA | BC151273.1 | - | 3 - 4 | 1678 | 73 | |
More Information
Set Membership: |
|

ADAMTS1
Genomic Map