Assay Details
| Gene Symbol: MRPL51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101088.1 | NP_001094558.1 | 1 - 2 | 187 | 60 | |
| GenBank mRNA | BC151349.1 | - | 1 - 2 | 187 | 60 | |
More Information
Set Membership: |
|

MRPL51
Genomic Map