Assay Details
| Gene Symbol: GPX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101113.2 | NP_001094583.1 | 1 - 2 | 203 | 91 | |
| GenBank mRNA | BC148128.1 | - | 1 - 2 | 150 | 91 | |
| CK836370.1 | - | 1 - 2 | 198 | 91 | ||
More Information
Set Membership: |
|

GPX7
Genomic Map