Assay Details
| Gene Symbol: RUBCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101897.2 | NP_001095367.2 | 5 - 6 | 747 | 58 | |
| GenBank mRNA | BC151465.1 | - | 5 - 6 | 742 | 58 | |
More Information
Set Membership: |
|

RUBCN
Genomic Map