Assay Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101901.2 | NP_001095371.1 | 6 - 7 | 874 | 65 | |
| GenBank mRNA | BC149413.1 | - | 6 - 7 | 875 | 65 | |
More Information
Set Membership: |
|

SLC22A31
Genomic Map