Assay Details
| Gene Symbol: CDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101947.1 | NP_001095417.1 | 1 - 2 | 525 | 93 | |
| GenBank mRNA | BC148005.1 | - | 1 - 2 | 525 | 93 | |
More Information
Set Membership: |
|

CDX1
Genomic Map