Assay Details
| Gene Symbol: TBC1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101981.1 | NP_001095451.1 | 6 - 7 | 1377 | 66 | |
| GenBank mRNA | BC149120.1 | - | 6 - 7 | 1377 | 66 | |
More Information
Set Membership: |
|

TBC1D2
Genomic Map