Assay Details
| Gene Symbol: KBTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102013.1 | NP_001095483.1 | 1 | 2350 | 91 | |
| GenBank mRNA | BC151340.1 | - | 1 | 2350 | 91 | |
More Information
Set Membership: |
|

KBTBD2
Genomic Map