Assay Details
| Gene Symbol: CHST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102079.1 | NP_001095549.1 | 2 - 3 | 184 | 66 | |
| GenBank mRNA | BC148145.1 | - | 2 - 3 | 184 | 66 | |
More Information
Set Membership: |
|

CHST13
Genomic Map