Assay Details
| Gene Symbol: IMPACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102501.1 | NP_001095971.1 | 9 - 10 | 812 | 63 | |
| GenBank mRNA | BC151281.1 | - | 9 - 10 | 812 | 63 | |
More Information
Set Membership: |
|

IMPACT
Genomic Map