Assay Details
| Gene Symbol: FAM120B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102507.1 | NP_001095977.1 | 4 - 5 | 1544 | 59 | |
| GenBank mRNA | BC148988.1 | - | 4 - 5 | 1544 | 59 | |
More Information
Set Membership: |
|

FAM120B
Genomic Map