Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103230.1 | NP_001096700.1 | 3 - 4 | 976 | 78 | |
| GenBank mRNA | BC148033.1 | - | 3 - 4 | 976 | 78 | |
More Information
Set Membership: |
|

VWA1
Genomic Map