Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145791.1 | NP_001139263.1 | 4 - 5 | 361 | 91 | |
| NM_001145792.1 | NP_001139264.1 | 4 - 5 | 361 | 91 | ||
| GenBank mRNA | DT849463.1 | - | 1 - 2 | 91 | 91 | |
| EE218140.1 | - | 4 - 5 | 356 | 91 | ||
| FE007848.1 | - | 4 - 5 | 348 | 91 | ||
| Gene Symbol: LOC100271850-MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027315.1 | 4 - 5 | 361 | 91 | ||
More Information
Set Membership: |
|

BORCS8
Genomic Map