Assay Details
| Gene Symbol: CARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103249.1 | NP_001096719.1 | 19 - 20 | 2157 | 59 | |
| GenBank mRNA | BC114705.1 | - | 19 - 20 | 2157 | 59 | |
More Information
Set Membership: |
|

CARS1
Genomic Map