Assay Details
| Gene Symbol: SNX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103257.1 | NP_001096727.1 | 1 - 2 | 198 | 73 | |
| GenBank mRNA | BC150127.1 | - | 1 - 2 | 198 | 73 | |
More Information
Set Membership: |
|

SNX12
Genomic Map