Assay Details
| Gene Symbol: ABCG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105343.1 | NP_001098813.1 | 5 - 6 | 763 | 56 | |
| GenBank mRNA | BC153242.1 | - | 5 - 6 | 763 | 56 | |
More Information
Set Membership: |
|

ABCG4
Genomic Map