Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105353.1 | NP_001098823.1 | 1 - 2 | 88 | 135 | |
| GenBank mRNA | BC147951.1 | - | 1 - 2 | 88 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC147951.1, NM_001105353.1 transcripts.
Set Membership: |
|

ZNF500
Genomic Map