Assay Details
| Gene Symbol: PTS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113537.1 | NP_001107009.1 | 2 - 3 | 210 | 64 | |
| GenBank mRNA | CB455463.1 | - | 2 - 3 | 215 | 64 | |
More Information
Set Membership: |
|

PTS
Genomic Map