Assay Details
| Gene Symbol: TFF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114509.2 | NP_001107981.1 | 2 - 3 | 272 | 151 | |
| GenBank mRNA | BC102088.1 | - | 2 - 3 | 273 | 151 | |
More Information
Set Membership: |
|

TFF3
Genomic Map