Assay Details
| Gene Symbol: LAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034570.2 | NP_001029742.1 | 5 - 6 | 870 | 68 | |
| NM_001113244.1 | NP_001106715.1 | 5 - 6 | 870 | 68 | ||
| GenBank mRNA | BC102819.1 | - | 5 - 6 | 852 | 68 | |
| DT826059.1 | - | 3 - 4 | 392 | 68 | ||
| DV891215.1 | - | 5 - 6 | 715 | 68 | ||
More Information
Set Membership: |
|

LAMP2
Genomic Map