Assay Details
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 7 - 8 | 657 | 95 | |
GenBank mRNA | BC105446.1 | - | 7 - 8 | 693 | 95 | |
DV883546.1 | - | 4 - 5 | 339 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
