Assay Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078099.2 | NP_001071567.2 | 3 - 4 | 670 | 72 | |
| GenBank mRNA | BC123671.1 | - | 3 - 4 | 689 | 72 | |
| BT030476.1 | - | 3 - 4 | 670 | 72 | ||
More Information
Set Membership: |
|

DPH2
Genomic Map