Assay Details
| Gene Symbol: ST8SIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098113.1 | NP_001091582.1 | 3 - 4 | 269 | 67 | |
| GenBank mRNA | AM422133.1 | - | 3 - 4 | 269 | 67 | |
More Information
Set Membership: |
|

ST8SIA2
Genomic Map