Assay Details
| Gene Symbol: MRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098118.1 | NP_001091587.1 | 4 - 5 | 528 | 67 | |
| GenBank mRNA | BC140603.1 | - | 4 - 5 | 528 | 67 | |
| BC142259.1 | - | 4 - 5 | 625 | 67 | ||
| BC142462.1 | - | 4 - 5 | 482 | 67 | ||
More Information
Set Membership: |
|

MRAS
Genomic Map