Assay Details
| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098141.1 | NP_001091610.1 | 3 - 4 | 179 | 80 | |
| GenBank mRNA | AB300617.1 | - | 3 - 4 | 179 | 80 | |
More Information
Set Membership: |
|

GYPA
Genomic Map