Assay Details
| Gene Symbol: TWNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098463.1 | NP_001091933.1 | 3 - 4 | 2158 | 83 | |
| GenBank mRNA | BC140487.1 | - | 3 - 4 | 2158 | 83 | |
More Information
Set Membership: |
|

TWNK
Genomic Map