Assay Details
| Gene Symbol: DDX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099045.1 | NP_001092515.1 | 4 - 5 | 1451 | 60 | |
| GenBank mRNA | BC146184.1 | - | 4 - 5 | 1451 | 60 | |
More Information
Set Membership: |
|

DDX24
Genomic Map